missing translation for 'onlineSavingsMsg'
Learn More

Melford NAD

Product Code. p-200087198
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 30018577

Brand: Melford N301101.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

β-Nicotinamide-Adenine Dinucleotide, Oxidized Form (β-NAD) is suitable for Enzyme synthesis / Biocatalysis applications.

β-Nicotinamide-Adenine Dinucleotide, Oxidized Form (β-NAD) is suitable for Enzyme synthesis / Biocatalysis applications. It is also applicable for making Enzyme Multiplied Immunoassay Technique (EMIT), Therapeutic Drug monitoring (TDM) and Blood Glucose reagents.

Specifications

Quantity 1 g
Molecular Formula C21H27N7O14P2 • XH2O
CAS 53-84-9
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.