Learn More
Specac™ NaCl Sealed Heatable Liquid Flow Cell VTCH Incompatible
Product Code. 15200228
Encompass_Preferred
Change view
Click to view available options
Type:
NaCl Sealed Heatable Liquid Flow Cell
Product Code. Type
15200228 NaCl Sealed Heatable Liquid Flow Cell
1 options
Product Code. 15200228 Supplier Specac™ Supplier No. GS20570
This item requires a price update prior to being ordered.
Request a Price for this item here.
This cell is designed for use in Specac heating jackets GS20710 and GS20730. This is a sealed heatable liquid flow cell with NaCl windows and choice of pathlength from 0.025mm up to 1.0mm. This cell is fitted with 1/16in. Swagelok fittings for flow purposes.
Specifications
Type NaCl Sealed Heatable Liquid Flow Cell
Description Pathlength from 0.025 mm up to 1.0 mm