Learn More
Specac™ NaCl Demountable Heatable Liquid Static Cell
Description
This heatable liquid cell is designed for high sensitivity ATR measurements with FTIR or dispersive instruments. This is a demountable cell with NaCl windows and Mylar Spacer that is available with a user defined pathlength ranging from 0.006mm up to 1.0mm.
Specifications
Specifications
| Type | NaCl Demountable Heatable Liquid Static Cell |
| Description | Pathlengths available ranging from 0.006 mm up to 1.0 mm |