Learn More
Specac™ NaCl Demountable Heatable Flow Cell VTCH Incompatible
Product Code. 15250228
Encompass_Preferred
Change view
Click to view available options
Type:
NaCl Demountable Heatable Flow Cell
Product Code. Type
15250228 NaCl Demountable Heatable Flow Cell
1 options
Product Code. 15250228 Supplier Specac™ Supplier No. GS20580
Only null left
Add to basket
Add to basket
This cell is designed for use in Specac heating jackets GS20710 and GS20730. This is a demountable heatable liquid flow cell with NaCl windows and choice of pathlength from 0.025mm up to 1.0mm. This cell is fitted with 1/16in. Swagelok fittings for flow purposes.
Specifications
Type NaCl Demountable Heatable Flow Cell
Description Pathlength from 0.025 mm up to 1.0 mm