missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NACC2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA561624

Product Code. 15789431

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RVRKRWLPKI KSMLPEGVEM YRTVMGSAAA SVPLDPEFPP AAAQVFEQRI YAERRGDAAT IVALRTDAVN VDLSAAAN Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Functions as a transcriptional repressor through its association with the NuRD complex. Recruits the NuRD complex to the promoter of MDM2, leading to the repression of MDM2 transcription and subsequent stability of p53/TP53. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NACC2
Polyclonal
Unconjugated
Nacc2
0610020I02Rik; AI448087; BEN domain containing 9; BEND9; BTB (POZ) domain containing 14A; BTB/POZ domain-containing protein 14A; BTBD14; Btbd14a; BTBD31; C030048H19Rik; MGC23427; NAC2; NAC-2; NACC family member 2; NACC family member 2, BEN and BTB (POZ) domain containing; Nacc2; nucleus accumbens associated 2, BEN and BTB (POZ) domain containing; nucleus accumbens-associated protein 2; RBB; repressor with BTB domain and BEN domain; transcription repressor with a BTB domain and a BEN domain
Rabbit
Antigen affinity chromatography
RUO
138151
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96BF6
Nacc2
Recombinant protein corresponding to Human NACC2. Recombinant protein control fragment (Product #RP-100971).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato