missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NACC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562709
This item is not returnable.
View return policy
Description
Immunogen sequence: DEEDDEAYDT MVEEQYGQMY IKASGSYAVQ EKPEPVPLES RSCVLIRRDL VALPA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
Functions as a transcriptional repressor through its association with the NuRD complex. Recruits the NuRD complex to the promoter of MDM2, leading to the repression of MDM2 transcription and subsequent stability of p53/TP53. [UniProt].
Specifications
| NACC2 | |
| Polyclonal | |
| Unconjugated | |
| Nacc2 | |
| 0610020I02Rik; AI448087; BEN domain containing 9; BEND9; BTB (POZ) domain containing 14A; BTB/POZ domain-containing protein 14A; BTBD14; Btbd14a; BTBD31; C030048H19Rik; MGC23427; NAC2; NAC-2; NACC family member 2; NACC family member 2, BEN and BTB (POZ) domain containing; Nacc2; nucleus accumbens associated 2, BEN and BTB (POZ) domain containing; nucleus accumbens-associated protein 2; RBB; repressor with BTB domain and BEN domain; transcription repressor with a BTB domain and a BEN domain | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 138151 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96BF6 | |
| Nacc2 | |
| Recombinant protein corresponding to Human NACC2. Recombinant protein control fragment (Product #RP-100970). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction