Learn More
Abnova™ NACAP3/CEN12q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0174.200uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16003275
  • £2030.00 / 200µL
Estimated Shipment: 10-06-2024
to see stock.
Includes: DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Specifications
Store at 4°C in the dark.
Tube
101928685
200 μL
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
FISH-Ce
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe