missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NACAD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101691
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NACAD.
May prevent inappropriate targeting of non-secretory polypeptides to the endoplasmic reticulum (ER). May bind to nascent polypeptide chains as they emerge from the ribosome and block their interaction with the signal recognition particle (SRP), which normally targets nascent secretory peptides to the ER. May also reduce the inherent affinity of ribosomes for protein translocation sites in the ER membrane (M sites).
Specifications
| NACAD | |
| Polyclonal | |
| Unconjugated | |
| NACAD | |
| D230024G13Rik; Kiaa0363; mKIAA0363; NAC alpha domain containing; NACAD; NAC-alpha domain-containing protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23148 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15069 | |
| NACAD | |
| A synthesized peptide derived from human NACAD(Accession O15069), corresponding to amino acid residues E41-P91. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction