missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NACA1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33336-20ul
This item is not returnable.
View return policy
Description
NACA1 Monoclonal antibody specifically detects NACA1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| NACA1 | |
| Monoclonal | |
| Western Blot 1:2000 - 1:20000, ELISA Recommended starting concentration is 1 μg/mL | |
| Allergen Hom s 2, Alpha-NAC, MGC117224, NACA1, NAC-alpha, nascent polypeptide-associated complex alpha subunit, nascent polypeptide-associated complex subunit alpha, nascent-polypeptide-associated complex alpha polypeptide | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-215 of human NACA1 (NP_005585.1).,, Sequence:, MPGEATETVPATEQELPQPQAETGSGTESDSDESVPELEEQDSTQATTQQAQLAAAAEIDEEPVSKAKQSRSEKKARKAMSKLGLRQVTGVTRVTIRKSKNILFVITKPDVYKSPASDTYIVFGEAKIEDLSQQAQLAAAEKFKVQGEAVSNIQENTQTPTVQEESEEEEVDETGVEVKDIELVMSQANVSRAKAVRALKNNSNDIVNAIMELTM | |
| 20 μL | |
| DNA replication Transcription Translation and Splicing | |
| 4666 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction