missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584538
This item is not returnable.
View return policy
Description
Immunogen sequence: RVVRKSWMPK VKVLKAEDDA YTTFISETGK IEPDMMGVEH GFETASHEGE AGPSAEAL Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 73%.
Members of the BTB/POZ family of transcriptional regulators, including BTBD14B, contain a conserved motif in the N-terminal region critical for protein-protein interactions and assembly of high molecular mass complexes (Korutla et al., 2002 [PubMed 11906783]).
Specifications
| NAC1 | |
| Polyclonal | |
| Unconjugated | |
| NACC1 | |
| 2010001H03Rik; 4930511N13Rik; BEN domain containing 8; BEND8; BTB (POZ) domain containing 14B; BTB/POZ domain-containing protein 14B; Btbd14b; BTBD30; FLJ37383; Nac1; NAC-1; Nacc1; nucleus accumbens associated 1; nucleus accumbens associated 1, BEN and BTB (POZ) domain containing; nucleus accumbens-1; nucleus accumbens-associated protein 1; nucleus accumbens-associated protein 1; LOW QUALITY PROTEIN: nucleus accumbens-associated protein 1; transcriptional repressor NAC1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 112939 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96RE7 | |
| NACC1 | |
| Recombinant protein corresponding to Human NACC1. Recombinant protein control fragment (Product #RP-104240). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction