missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAC1 Antibody (PCRP-NACC1-1A8), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08833CL1
This item is not returnable.
View return policy
Description
NAC1 Monoclonal antibody specifically detects NAC1 in Human samples. It is validated for Flow Cytometry,Protein Array
Specifications
| NAC1 | |
| Monoclonal | |
| CoraFluor 1 | |
| BEND8, BTB (POZ) domain containing 14B, BTB/POZ domain-containing protein 14B, BTBD14B, NAC1BEN domain containing 8, NAC-1FLJ37383, nucleus accumbens associated 1, BEN and BTB (POZ) domain containing, nucleus accumbens-associated protein 1, transcriptional repressor NAC1 | |
| Recombinant full-length human NAC1 protein (Uniprot: Q96RE7) | |
| 0.1 mL | |
| Cancer, Chromatin Research, Stem Cell Markers, Transcription Factors and Regulators | |
| 112939 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Protein Array | |
| PCRP-NACC1-1A8 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction