missing translation for 'onlineSavingsMsg'
Learn More

NAC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£169.00 - £255.00

Specifications

Antigen NAC1
Dilution Immunocytochemistry/ Immunofluorescence 1:300-1:10000
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18621852
View Documents
Novus Biologicals
NBP2-98709-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18633842
View Documents
Novus Biologicals
NBP2-98709-50ul
50 μg
£169.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NAC1 Polyclonal antibody specifically detects NAC1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

NAC1
Immunofluorescence
Unconjugated
Rabbit
Cancer, Chromatin Research, Stem Cell Markers, Transcription Factors and Regulators
0.2 um filtered solution in PBS
112939
IgG
Antigen and protein A Affinity-purified
Immunocytochemistry/ Immunofluorescence 1:300-1:10000
Polyclonal
Purified
RUO
Human
BEND8, BTB (POZ) domain containing 14B, BTB/POZ domain-containing protein 14B, BTBD14B, NAC1BEN domain containing 8, NAC-1FLJ37383, nucleus accumbens associated 1, BEN and BTB (POZ) domain containing, nucleus accumbens-associated protein 1, transcriptional repressor NAC1
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NAC1.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.