missing translation for 'onlineSavingsMsg'
Learn More

Nabumetone

Product Code. 15495339
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495339 5 g 5g
15405349 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495339 Supplier Thermo Scientific Alfa Aesar Supplier No. J63072.06
  • ONLINE EXCLUSIVE PRICE
    £109.14
    £87.65 / 5g
    Save £21.49
    19% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42924-53-8 | C15H16O2 | 228.291 g/mol

Anti-inflammatory, anti-bacterial. A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Anti-inflammatory, anti-bacterial
  • A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42924-53-8
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.291
MDL Number MFCD00079518
InChI Key BLXXJMDCKKHMKV-UHFFFAOYSA-N
Synonym nabumetone, relafen, 4-6-methoxynaphthalen-2-yl butan-2-one, arthaxan, nabumetona, relifex, listran, relifen, nabumeton, consolan
PubChem CID 4409
ChEBI CHEBI:7443
IUPAC Name 4-(6-methoxynaphthalen-2-yl)butan-2-one
SMILES CC(=O)CCC1=CC2=C(C=C1)C=C(C=C2)OC

Specifications

Melting Point 76°C to 78°C
Color White
Quantity 5 g
Merck Index 14,6343
Solubility Information Soluble in alcohol or chloroform
Formula Weight 228.29
Physical Form Crystalline Powder
Chemical Name or Material Nabumetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.