missing translation for 'onlineSavingsMsg'
Learn More

Nabumetone, Mikromol™

Product Code. 17973566
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973566 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17973566 Supplier Mikromol Supplier No. MM0153.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Nabumetone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nabumetone
CAS 42924-53-8
Analyte or Component Names Nabumetone
Shipping Condition Room Temperature
Molecular Formula C15 H16 O2
Certifications/Compliance ISO 17034
Synonym Nabumetone, 4-(6-Methoxy-2-naphthalenyl)-2-butanone, 1-(2'-Methoxynaphth-6'-yl)butan-3-one, 4-(6-Methoxy-2-naphthyl)-2-butanone, Arthaxan, BRL 14777, Balmox, Consolan, Nabumeton, Nabumetone, Nabuser, Relafen, Relifen, Relifex, BRL-14777
InChI Formula InChI=1S/C15H16O2/c1-11(16)3-4-12-5-6-14-10-15(17-2)8-7-13(14)9-12/h5-10H,3-4H2,1-2H3
SMILES COc1ccc2cc(CCC(=O)C)ccc2c1
IUPAC Name 4-(6-methoxynaphthalen-2-yl)butan-2-one
Molecular Weight (g/mol) 228.29
Quantity 500 mg
Formula Weight 228.115 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.