missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAB2 Monoclonal Antibody (OTI1E12)
GREENER_CHOICE

Product Code. 15986234
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15986234 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15986234 Supplier Invitrogen™ Supplier No. MA526644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NAB2 Monoclonal Antibody for Western Blot, IHC (P)

Transcriptional control is in part regulated by interactions between DNA-bound transcription factors, such as Egr-1/NGFI-A, and coregulatory proteins, such as NAB (for NGFI-A-binding proteins). The evolutionarily conserved NAB proteins, NAB1 and NAB2, are corepressors of EGF-1/NGFI-A. Both NAB1 and NAB2 contain an amino terminal NAB conserved domain 1 (NCB1), which is required for binding NGFI-A, and a carboxy terminal NCD2 domain, which is responsible for the repressor function of NAB proteins. NAB2 is principally localized in the nucleus and may play a role in the downregulation of NGFI-A activity as well as in controlling fundamental processes such as cell division, differentiation and apoptosis. NAB2 has a predicted molecular mass of 56 kDa and localizes to chromosome 12q13.3-14.1, a region that is rearranged in several solid tumors, lipomas and liposarcomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAB2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1E12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NAB2
Gene Accession No. Q15742
Gene Alias AI451907; EGR1 binding protein 2; EGR-1-binding protein 2; MADER; Melanoma-associated delayed early response protein; NAB2; NGFI-A binding protein 2; NGFI-A binding protein 2 (EGR1 binding protein 2); NGFI-A binding protein 2 (EGR-1 binding protein 2); NGFI-A-binding protein 2; Protein MADER
Gene Symbols NAB2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 178-442 of NAB2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4665
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.