missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Nab2 Monoclonal antibody specifically detects Nab2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | Nab2 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI5D7 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EGR-1-binding protein 2, MADEREGR1 binding protein 2, Melanoma-associated delayed early response protein, MGC75085, NGFI-A binding protein 2 (EGR1 binding protein 2), NGFI-A-binding protein 2, Protein MADER |
| Host Species | Mouse |
| Immunogen | Recombinant protein fragment corresponding to amino acids 178-442 of human Nab2 (NP_005958) produced in E.coli. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?