missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nab2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38193-100ul
This item is not returnable.
View return policy
Description
Nab2 Polyclonal antibody specifically detects Nab2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Nab2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EGR-1-binding protein 2, MADEREGR1 binding protein 2, Melanoma-associated delayed early response protein, MGC75085, NGFI-A binding protein 2 (EGR1 binding protein 2), NGFI-A-binding protein 2, Protein MADER | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human Nab2 (NP_005958.1).,, Sequence:, MHRAPSPTAEQPPGGGDSARRTLQPRLKPSARAMALPRTLGELQLYRVLQRANLLSYYETFIQQGGDDVQQLCEAGEEEFLEIMALVGMATKPLHVRRLQKALREWATNPGLFSQPVPAVPVSSIPLFKISETAGTRKGSMSNGHGSPGEKAGSARSFSPKSPLELGEKLSPLPGGPGAGDPRIWPGRSTPESDVGAGGE | |
| 100 μL | |
| Neuroscience, Transcription Factors and Regulators | |
| 4665 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction