missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Nab2 Antibody (4H6), Novus Biologicals™

Product Code. 18344348 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344348 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344348 Supplier Novus Biologicals™ Supplier No. H00004665M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Nab2 Monoclonal antibody specifically detects Nab2 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nab2
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 4H6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005958
Gene Alias EGR-1-binding protein 2, MADEREGR1 binding protein 2, Melanoma-associated delayed early response protein, MGC75085, NGFI-A binding protein 2 (EGR1 binding protein 2), NGFI-A-binding protein 2, Protein MADER
Host Species Mouse
Immunogen NAB2 (NP_005958.1, 421 a.a. ∽ 525 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DGHLQAVGSCPRLTPPPADLPLALPAHGLWSRHILQQTLMDEGLRLARLVSHDRVGRLSPCVPAKPPLAEFEEGLLDRCPAPGPHPALVEGRRSSVKVEAEASRQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4665
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.