missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18541-1-AP-20UL
This item is not returnable.
View return policy
Description
Acts as a transcriptional repressor for zinc finger transcription factors EGR1 and EGR2.Specifications
| NAB1 | |
| Polyclonal | |
| Unconjugated | |
| NAB1 | |
| EGR 1 binding protein 1, NAB1, NGFI A binding protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17936, 4664, 64824 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13506, Q61122, Q62722 | |
| Nab1 | |
| NAB1 Fusion Protein Ag13407 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction