missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAALADase-like 1/NAALADL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£199.00 - £353.00
Specifications
| Antigen | NAALADase-like 1/NAALADL1 |
|---|---|
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616195
|
Novus Biologicals
NBP2-99724-100ul |
100 μg |
£353.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18634895
|
Novus Biologicals
NBP2-99724-50ul |
50 μg |
£199.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NAALADase-like 1/NAALADL1 Polyclonal antibody specifically detects NAALADase-like 1/NAALADL1 in Human samples. It is validated for ELISASpecifications
| NAALADase-like 1/NAALADL1 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| I100, NAALADASEL, NAALADL1 N-acetylated alpha-linked acidic dipeptidase-like 1 | |
| Produced in rabbits immunized with purified, recombinant Human NAALADase-like 1/NAALADL1 (Accession#: NP_005459.2; Pro 29-Leu 740) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 10004 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title