missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAALADase-like 1/NAALADL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | NAALADase-like 1/NAALADL1 |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18436081
|
Novus Biologicals
NBP2-14584-25ul |
25ul |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18158291
|
Novus Biologicals
NBP2-14584 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NAALADase-like 1/NAALADL1 Polyclonal specifically detects NAALADase-like 1/NAALADL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NAALADase-like 1/NAALADL1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 10004 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: RLSDSFFLPLKVSDYSETLRSFLQAAQQDLGALLEQHSISLGPLVTAVEKFEAEAAALGQRISTLQKGSPDPLQVRMLN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| I100, NAALADASEL, NAALADL1 N-acetylated alpha-linked acidic dipeptidase-like 1 | |
| NAALADL1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title