missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAALADase-like 1/NAALADL1 Antibody (026), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
NAALADase-like 1/NAALADL1 Monoclonal antibody specifically detects NAALADase-like 1/NAALADL1 in Mouse samples. It is validated for ELISA
Specifications
Specifications
| Antigen | NAALADase-like 1/NAALADL1 |
| Clone | 026 |
| Host Species | Rabbit |
| Gene ID (Entrez) | 10004 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?