Learn More
NAALADase-like 1/NAALADL1 Antibody (005), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90747MFV610
Product Code. 18491319
  • £492.00 / 0.10mL
Description
NAALADase-like 1/NAALADL1 Monoclonal antibody specifically detects NAALADase-like 1/NAALADL1 in Mouse samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.1 mL