missing translation for 'onlineSavingsMsg'
Learn More

NAALADase-2/NAALAD2 Antibody (756030) [DyLight 680], Novus Biologicals™

Product Code. 30733192 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733192 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733192 Supplier Novus Biologicals Supplier No. FAB7658Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NAALADase-2/NAALAD2 Monoclonal antibody specifically detects NAALADase-2/NAALAD2 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAALADase-2/NAALAD2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 756030
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias GCPIII, glutamate carboxypeptidase III, GPCIII, MGC116996, MGC26353, NAADALASE2, NAALADase II, N-acetylated alpha-linked acidic dipeptidase 2, N-acetylated alpha-linked acidic dipeptidase II, N-acetylated-alpha-linked acidic dipeptidase 2, N-acetylated-alpha-linked acidic dipeptidase II
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human NAALADase-2/NAALAD2, Lys32-Leu740, Accession # Q9Y3Q0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10003
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.