missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAA25 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £366.00
Specifications
| Antigen | NAA25 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619721
|
Novus Biologicals
NBP2-94737-0.02ml |
0.02 mL |
£159.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689821
|
Novus Biologicals
NBP2-94737-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NAA25 Polyclonal antibody specifically detects NAA25 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| NAA25 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology | |
| PBS (pH 7.3), 50% glycerol | |
| 80018 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| C12orf30DKFZp667K2112, chromosome 12 open reading frame 30, FLJ13089, MDM20mitochondrial distribution and morphology 20, Mitochondrial distribution and morphology protein 20, N(alpha)-acetyltransferase 25, NatB auxiliary subunit, N-alpha-acetyltransferase 25, NatB auxiliary subunit, NAP1, NatB complex subunit MDM20, N-terminal acetyltransferase B complex subunit MDM20, N-terminal acetyltransferase B complex subunit NAA25, p120 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 703-972 of human NAA25 (NP_079229.2). LNHPVEPKNSEKTAENGVSSRIDILRLLLQQLEATLETGKRFIEKDIQYPFLGPVPTRMGGFFNSGCSQCQISSFYLVNDIYELDTSGLEDTMEIQERIENSFKSLLDQLKDVFSKCKGDLLEVKDGNLKTHPTLLENLVFFVETISVILWVSSYCESVLRPYKLNLQKKKKKKKETSIIMPPVFTSFQDYVTGLQTLISNVVDHIKGLETHLIALKLEELILEDTSLSPEERKFSKTVQGKVQSSYLHSLLEMGELLKKRLETTKKLKI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title