missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAA11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557462
This item is not returnable.
View return policy
Description
Immunogen sequence: DELRRQMDLK KGGYVVLGSR ENQETQGSTL SDSEEACQQK NPATEESGSD SKEPKESVES TNVQDSSE Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 63%.
Displays alpha (N-terminal) acetyltransferase activity. Proposed alternative catalytic subunit of the N-terminal acetyltransferase A (NatA) complex.
Specifications
| NAA11 | |
| Polyclonal | |
| Unconjugated | |
| NAA11 | |
| NAA11 | |
| Recombinant protein corresponding to Human NAA11. Recombinant protein control fragment (Product #RP-96697). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BSU3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84779 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction