missing translation for 'onlineSavingsMsg'
Learn More

Na-Cbz-L-tryptophan, TRC

Product Code. 30353309
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30353309 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353309 Supplier Toronto Research Chemicals Supplier No. TRCC227795250G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nalpha-Cbz-L-tryptophan
CAS 7432-21-5
Molecular Formula C19H18N2O4
Quantity 250 g
Synonym N-[(Phenylmethoxy)carbonyl]-L-tryptophan, N-Carboxy-L-tryptophan N-Benzyl Ester, (S)-2-(((Benzyloxy)carbonyl)amino)-3-(1H-indol-3-yl)propanoic Acid, Benzyloxycarbonyl-L-tryptophan, Carbobenzoxy-L-tryptophan, L-(Carbobenzyloxy)tryptophan, N-(Benzyloxycarbonyl)tryptophan, N-Benzyloxycarbonyl-L-tryptophan, N-Carbobenzoxy-L-tryptophan, N-Carbobenzyloxy-L-tryptophan, N-Cbz-L-Tryptophan, NSC 521831, Nalpha-Benzyloxycarbonyl-L-tryptophan
InChI Formula InChI=1S/C19H18N2O4/c22-18(23)17(10-14-11-20-16-9-5-4-8-15(14)16)21-19(24)25-12-13-6-2-1-3-7-13/h1-9,11,17,20H,10,12H2,(H,21,24)(H,22,23)/t17-/m0/s1
SMILES OC(=O)[C@H](Cc1c[nH]c2ccccc12)NC(=O)OCc3ccccc3
IUPAC Name (2S)-3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
Molecular Weight (g/mol) 338.36
Formula Weight 338.1267
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.