missing translation for 'onlineSavingsMsg'
Learn More

N7-(2-Hydroxyethyl)guanine-d4, TRC

Product Code. 30398010
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398010 25 mg -
30401048 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30398010 Supplier Toronto Research Chemicals Supplier No. TRCH94220225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246818-81-4
Molecular Formula C7H5D4N5O2
Molecular Weight (g/mol) 199.2
Synonym 2-Amino-1,7-dihydro-7-(2-hydroxyethyl-d4)-6H-purin-6-one, 7-(2-Hydroxyethyl-d4)guanine, 7-(β-Hydroxyethyl-d4)guanine, NSC 637506-d4
SMILES O=C1C2=C(N=CN2C([2H])([2H])C([2H])([2H])O)N=C(N)N1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C7H9N5O2/c8-7-10-5-4(6(14)11-7)12(1-2-13)3-9-5/h3,13H,1-2H2,(H3,8,10,11,14)/i1D2,2D2
Formula Weight 199.1007
Chemical Name or Material N7-(2-Hydroxyethyl)guanine-d4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.