missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6AMT2, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00221143-B03.50uL
This item is not returnable.
View return policy
Description
Sequence: MSDLEDDETPQLSAHALAALQEFYAEQKQQIEPGEDDKYNIGIIEENWQLSQFWYSQETALQLAQEAIAAVGEGGRIACVSAPSVYQKLRELCRENFSIYIFEYDKRFAMYGEEFIFYDYNNPLDLPERIAAHSFDIVIADPPYLSEECLRKTSETVKYLTRGKILLCTGAIMEEQAAELLGVKMCTFVPRHTRNLANEFRCYVNYDSGLDCGISpecifications
| N6AMT2 | |
| Polyclonal | |
| No additive | |
| NM_174928 | |
| N6AMT2 | |
| LOC221143 (NP_777588.1, 1 a.a. ∼ 214 a.a) full-length human protein. | |
| RUO | |
| 221143 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| N-6 adenine-specific DNA methyltransferase 2 (putative) | |
| ESP13 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction