missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6AMT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16211-1-AP-20UL
This item is not returnable.
View return policy
Description
Heterodimeric methyltransferase that catalyzes N5-methylation of ETF1 on ′Gln-185′, using S-adenosyl L-methionine as methyl donor. ETF1 needs to be complexed to ERF3 in its GTP-bound form to be efficiently methylated. May play a role in the modulation of arsenic-induced toxicity. May be involved in the conversion of monomethylarsonous acid (3+) into the less toxic dimethylarsonic acid.Specifications
| N6AMT1 | |
| Polyclonal | |
| Unconjugated | |
| N6AMT1 | |
| C21orf127, HEMK2, MTQ2, N6AMT, N6AMT1, PRED28 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288309, 29104, 67768 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| N6AMT1, Q9Y5N5 | |
| N6amt1 | |
| N6AMT1 Fusion Protein Ag9204 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction