missing translation for 'onlineSavingsMsg'
Learn More

N6-methyladenosine (m6A) Antibody, Novus Biologicals™

Product Code. 18319726 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319726 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319726 Supplier Novus Biologicals Supplier No. NBP318690

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

N6-methyladenosine (m6A) Polyclonal antibody specifically detects N6-methyladenosine (m6A) in Human, Mouse samples. It is validated for Dot Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen N6-methyladenosine (m6A)
Applications Dot Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.2 mg/ml
Conjugate Unconjugated
Dilution Dot Blot 1:400, RNA immunoprecipitation 1-2 ug/IP
Formulation PBS
Gene Alias (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against N6-methyladenosine (m6A) conjugated to LPH.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Epigenetics, metabolism
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.