missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-methyladenosine (m6A) Antibody (17-3-4-1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £320.00
Specifications
| Antigen | N6-methyladenosine (m6A) |
|---|---|
| Clone | 17-3-4-1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Dot Blot |
| Applications | Western Blot, Immunoprecipitation, Dot Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634139
|
Novus Biologicals
NBP2-50525 |
0.1 mL |
£320.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698456
|
Novus Biologicals
NBP2-50525-0.025ml |
0.025 mL |
£169.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N6-methyladenosine (m6A) Monoclonal antibody specifically detects N6-methyladenosine (m6A) in Human, Mouse, Yeast samples. It is validated for Western Blot, Immunoprecipitation, Dot BlotSpecifications
| N6-methyladenosine (m6A) | |
| 1 mg/mL | |
| Western Blot, Immunoprecipitation, Dot Blot | |
| Unconjugated | |
| Mouse | |
| Epigenetics, metabolism | |
| PBS | |
| Hapten N6-methyladenosine-5'-mono-phosphate conjugated to BSA of all N6-methyladenosine | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 17-3-4-1 | |
| Western Blot 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Dot Blot | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Yeast | |
| (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine | |
| IgG1 λ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title