missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-methyladenosine (m6A) Antibody (17-3-4-1), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50525AF532
This item is not returnable.
View return policy
Description
N6-methyladenosine (m6A) Monoclonal specifically detects N6-methyladenosine (m6A) in Human, Mouse, Yeast samples. It is validated for Western Blot.
Specifications
| N6-methyladenosine (m6A) | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein A purified | |
| Epigenetics, metabolism | |
| This antibody recognizes N6-methyladenosine in both modified RNA AND DNA. | |
| Store at 4°C in the dark. |
| Western Blot | |
| 17-3-4-1 | |
| Western Blot | |
| (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine | |
| Hapten N6-methyladenosine-5′-mono-phosphate conjugated to BSA of all N6-methyladenosine | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Yeast | |
| IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction