missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine, 98%

Product Code. 15430417
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15430417

Brand: Thermo Scientific Chemicals J65098.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C39H37N5O7

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methyladenosine
Color White to Yellow
Molecular Formula C39H37N5O7
MDL Number MFCD08704196
Quantity 1 g
Solubility Information Soluble in chloroform at 50mg/ml
Formula Weight 687.74
Percent Purity 98%
Physical Form Powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.