missing translation for 'onlineSavingsMsg'
Learn More

N5-(1-Iminoethyl) L-Ornithine Hydrochloride, TRC

Product Code. 30400989
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400989 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400989 Supplier Toronto Research Chemicals Supplier No. TRCI415000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 150403-88-6
Molecular Formula C7 H15 N3 O2 . Cl H
Quantity 100 mg
InChI Formula InChI=1S/C7H15N3O2.ClH/c1-5(8)10-4-2-3-6(9)7(11)12;/h6H,2-4,9H2,1H3,(H2,8,10)(H,11,12);1H/t6-;/m0./s1
SMILES Cl.CC(=N)NCCC[C@H](N)C(=O)O
IUPAC Name (2S)-2-amino-5-(ethanimidoylamino)pentanoic acid;hydrochloride
Molecular Weight (g/mol) 209.67
Formula Weight 209.0931
Chemical Name or Material L-N5-(1-Iminoethyl) Ornithine, Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.