Learn More
Invitrogen™ N4BP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117135
Description
Positive Samples: Jurkat, 293T, Mouse brain, Mouse pancreas Immunogen sequence: TQGEESLFET DDDVITVGDV RPPEYTQSAA TGPSSARDEV VVQEDSRNKA RTPVSELTKH MDTVFSSSPD VLFVPVNGLS PDEDALSKDR VCHKRRSSDT EERHTKKQFS LENVPEGELL PDGKGSAGNE VIDLSDPASN STNLSPDGKD TTEEMEYNIL VNFFKTMGYS QEIVEKVIRE YGPSTEPLLL LEEIEKENKR LQEDRDFPPC TVYPDASQSR NAGVGSTTNE LTADSTPKKA QSHTEQSMVE RFSQLPFKDS KHCTSNCKVN SFRTVPVGQK QEIWGSKQNS SCTVDLETDG HSASAASASP KDISFVSRGA SGHQQRNPAF PENGFQQQTE PLLPNNTKPA CEKRSGSCSS PQPKPNYPPL SPPLPLPQLL PSVTEARLGG SSDHIDSSVT G.
Nedd4-binding partner-1 (N4BP1) is a developmentally expressed protein interactor and monoubiquitylation substrate of Nedd4, a HECT domain-containing ubiquitin ligase that mediates ubiquitylation and proteasome degradation of target proteins and may have a fundamental role to play in embryonic processes. N4BP1 can undergo Nedd4-mediated polyubiquitylation and proteosomal degradation. N4BP1 also functions as a negative regulator of AIF4 (also known as ITCH or Atrophin-1), an E3-ubiquitin ligase, by interacting with the second WW domain of AIF4 and impairing ubiquitination of substrates.
Specifications
| N4BP1 | |
| Polyclonal | |
| Unconjugated | |
| N4BP1 | |
| AI481586; C81621; Kiaa0615; N4BP1; Nedd4 binding protein 1; NEDD4-binding protein 1; RGD1305179 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 80750, 9683 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.91 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.3 | |
| O75113, Q6A037 | |
| N4BP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 190-590 of mouse N4BP1 (NP_0850402). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.