missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N4BP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38551-100ul
This item is not returnable.
View return policy
Description
N4BP1 Polyclonal antibody specifically detects N4BP1 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| N4BP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| FLJ31821, KIAA0615, MGC176730, NEDD4 binding protein 1, NEDD4-binding protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 190-590 of mouse N4BP1 (NP_085040.2).,, Sequence:, TEARLGGSSDHIDSSVTGVQRFRDTLKIPYKLELKNEPGRADLKHIVIDGSNVAITHGLKKFFSCRGIAIAVEYFWKLGNRNITVFVPQWRTRRDPNITEQHFLTQLQELGILSLTPARMVFGERIASHDDRFLLHLADKTGGIIVTNDNFREFVTESVSWREIITKRLLQYTFVGDIFMVPDDPLGRNGPRLEEFLRKEAFLRHMQPLLNALPSVGTFDPGFRSPSTQVANNSHQPPPRIQTSSSPWLPQQSHFTALATLPSMQQNPPLPAQRSSAETSELREALLKIFPDSEQKL | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9683 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction