missing translation for 'onlineSavingsMsg'
Learn More

N4BP1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18423985 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423985 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423985 Supplier Novus Biologicals Supplier No. NBP176531JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

N4BP1 Polyclonal antibody specifically detects N4BP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen N4BP1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias FLJ31821, KIAA0615, MGC176730, NEDD4 binding protein 1, NEDD4-binding protein 1
Host Species Rabbit
Immunogen Antibody was raised against a 18 amino acid synthetic peptide near the amino terminus of human N4BP1. The immunogen is located within the first 50 amino acids of N4BP1. Amino Acid Squence: TAPAEKAELLEQSRGRIE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9683
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.