missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N4-Methylcytidine Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £359.00
Specifications
| Antigen | N4-Methylcytidine |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671028
|
Novus Biologicals
NBP3-05647-100ul |
100 μg |
£359.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619346
|
Novus Biologicals
NBP3-05647-20ul |
20 μg |
£149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N4-Methylcytidine Polyclonal antibody specifically detects N4-Methylcytidine in Human samples. It is validated for Western BlotSpecifications
| N4-Methylcytidine | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| 1-((2R,3S,4R,5S)-3,4-dihydroxy-5-(hydroxymethyl)-tetrahydrofuran-2-yl)-4-(methylamino)pyrimidin-2(1H)-one, 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-(methylamino)pyrimidin-2-one, 1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-(methylamino)pyrimidin-2-one, 10578-79-7, Cytidine, N-methyl-, N(4)-Methylcytidine, N-4-Methylcytidine, n-methylcytidine | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| Chemical compounds corresponding to N4-Methylcytidine/ 4mC. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title