missing translation for 'onlineSavingsMsg'
Learn More

N4-Hydroxycytosine (>90%), TRC

Product Code. 30403721
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403721 250 mg -
30404920 50 mg 50mg
30406308 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403721 Supplier Toronto Research Chemicals Supplier No. TRCH805078250MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20555-88-8
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.1
Synonym 6-(Hydroxyamino)-2(1H)-pyrimidinone
IUPAC Name 6-(hydroxyamino)-1H-pyrimidin-2-one
SMILES ONC1=CC=NC(=O)N1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C4H5N3O2/c8-4-5-2-1-3(6-4)7-9/h1-2,9H,(H2,5,6,7,8)
Formula Weight 127.0382
Chemical Name or Material N4-Hydroxycytosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.