missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N4-Benzoylcytidine, 99%

Product Code. 15467079
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
This item is not returnable. View return policy

Product Code. 15467079

Brand: Thermo Scientific Chemicals J65790.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13089-48-0 | C16H17N3O6 | 347.33 g/mol

N-Benzoylcytidine is a useful building block for oligoribonucleotide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoylcytidine is a useful building block for oligoribonucleotide synthesis.

Solubility
Soluble in 1N NaOH at 10mg/ml. Also soluble in methanol or DMSO

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13089-48-0
Molecular Formula C16H17N3O6
Molecular Weight (g/mol) 347.33
MDL Number MFCD00010572,MFCD01457402
InChI Key BNXBRFDWSPXODM-UHFFFAOYNA-N
Synonym n6-benzoylcytidine, n-1-2r,4r,5r-3,4-dihydroxy-5-hydroxymethyl oxolan-2-yl-2-oxopyrimidin-4-yl benzamide
PubChem CID 133611947
IUPAC Name N-[1-[(2S,5S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide
SMILES OCC1OC(C(O)C1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O

Specifications

Melting Point 230°C to 234°C (decomposition)
Color White
Quantity 5 g
Solubility Information Soluble in 1N NaOH at 10mg/ml; Also soluble in methanol or DMSO
Optical Rotation +55° (c=0.1 in Methanol)
Formula Weight 347.32
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N4-Benzoylcytidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.