missing translation for 'onlineSavingsMsg'
Learn More

N4-Acetylsulfadimidine (N4-Acetylsulfamethazine), Mikromol™

Product Code. 17992666
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17992666 25 mg 25mg
17971176 100 mg 100mg
30462246 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17992666 Supplier Mikromol Supplier No. MM1311.070025
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N4-Acetylsulfadimidine (N4-Acetylsulfamethazine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl Sulfamethazine
CAS 100-90-3
Analyte or Component Names N-Acetyl Sulfamethazine
Shipping Condition Room Temperature
Molecular Formula C14 H16 N4 O3 S
Certifications/Compliance ISO 9001
Synonym N-[4-[[(4,6-Dimethyl-2-pyrimidinyl)amino]sulfonyl]phenyl]acetamide, Acetanilide, 4'-[(4,6-dimethyl-2-pyrimidinyl)sulfamoyl]- (6CI,7CI,8CI), Acetanilide, p-(4,6-dimethyl-2-pyrimidylsulfamyl)- (4CI), 4'-[(4,6-Dimethyl-2-pyrimidinyl)sulfamoyl]acetanilide, Acetylsulfadimidine, Acetylsulfamethazine, N-Acetylsulfamethazine, N-[4-[[(4,6-Dimethyl-2-pyrimidinyl)amino]sulfonyl]phenyl]acetamide, N4-Acetylsulfadimidine, N4-Acetylsulfamethazine, N4-Acetylsulmet, Sulfamethazine-N4-acetyl
InChI Formula InChI=1S/C14H16N4O3S/c1-9-8-10(2)16-14(15-9)18-22(20,21)13-6-4-12(5-7-13)17-11(3)19/h4-8H,1-3H3,(H,17,19)(H,15,16,18)
SMILES CC(=O)Nc1ccc(cc1)S(=O)(=O)Nc2nc(C)cc(C)n2
IUPAC Name N-[4-[(4,6-dimethylpyrimidin-2-yl)sulfamoyl]phenyl]acetamide
Molecular Weight (g/mol) 320.37
Quantity 25 mg
Formula Weight 320.0943 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.