missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N4-Acetylcytosine, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
N4-Acetylcytosine
Specifications
Specifications
| Chemical Name or Material | N4-Acetylcytosine |
| CAS | 14631-20-0 |
| Analyte or Component Names | N4-Acetylcytosine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H7 N3 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Acetamide, N-(2,3-dihydro-2-oxo-4-pyrimidinyl)-, Acetamide, N-(1,2-dihydro-2-oxo-4-pyrimidinyl)- (8CI,9CI), Cytosine, N-acetyl- (6CI), N-(2,3-Dihydro-2-oxo-4-pyrimidinyl)acetamide, 2(1H)-Pyrimidinone, 4-(acetylamino)-, 4-N-Acetylcytosine, N-Acetylcytosine, NSC 210403 |
| InChI Formula | InChI=1S/C6H7N3O2/c1-4(10)8-5-2-3-7-6(11)9-5/h2-3H,1H3,(H2,7,8,9,10,11) |
| SMILES | CC(=O)NC1=CC=NC(=O)N1 |
| IUPAC Name | N-(2-oxo-1H-pyrimidin-6-yl)acetamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction