missing translation for 'onlineSavingsMsg'
Learn More

N4,5-Dimethyldeoxycytidine, TRC

Product Code. 30380361
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380361 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380361 Supplier Toronto Research Chemicals Supplier No. TRCD4681001MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25406-44-4
Molecular Formula C11H17N3O4
Quantity 1 mg
Synonym 2'-Deoxy-N,5-dimethyl-cytidine (6CI,7CI,8CI,9CI)
InChI Formula InChI=1S/C11H17N3O4/c1-6-4-14(11(17)13-10(6)12-2)9-3-7(16)8(5-15)18-9/h4,7-9,15-16H,3,5H2,1-2H3,(H,12,13,17)/t7-,8+,9+/m0/s1
SMILES CNC1=NC(=O)N(C=C1C)[C@H]2C[C@H](O)[C@@H](CO)O2
Molecular Weight (g/mol) 255.27
Formula Weight 255.1219
Chemical Name or Material N4,5-Dimethyldeoxycytidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.