missing translation for 'onlineSavingsMsg'
Learn More

N33-TEG-COOH, MedChemExpress

Product Code. 16495146
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495146 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16495146 Supplier Medchem Express Supplier No. HY108370
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress N33-TEG-COOH (N3-TEG-COOH) is a PEG-based PROTAC linker can be used in the synthesis of PROTACs.

N33-TEG-COOH (N3-TEG-COOH) is a PEG-based PROTAC linker can be used in the synthesis of PROTACs.

Specifications

Chemical Name or Material N33-TEG-COOH
CAS 201467-81-4
Color Colorless
Purity Grade Notes Research
Molecular Formula C10H19N3O6
Quantity 100 mg
Synonym N3-TEG-COOH 14-Azido-3,6,9,12-tetraoxatetradecanoic acid
Solubility Information Ethanol : 100 mg/mL (360.66 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (360.66 mM; Need ultrasonic)
SMILES O=C(O)COCCOCCOCCOCCN=[N+]=[N-]
Molecular Weight (g/mol) 277.27
Formula Weight 277.27
Percent Purity 96.0%
Grade Research
Physical Form Liquid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.