missing translation for 'onlineSavingsMsg'
Learn More

N3-Ethylthiosulfonylmethyl Lumiflavin, TRC

Product Code. 30383291
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383291 2.5 mg 2.50mg
30387262 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383291 Supplier Toronto Research Chemicals Supplier No. TRCE8180902.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N3-Ethylthiosulfonylmethyl Lumiflavin
Analyte or Component Names N3-Ethylthiosulfonylmethyl Lumiflavin
Shipping Condition Room Temperature
Molecular Formula C16H18N4O4S2
Synonym Lumiflavine N-Methanesulfonothioic Acid Ethyl Ester
InChI Formula InChI=1S/C16H18N4O4S2/c1-9-7-11-12(8-10(9)2)19(3)14-13(17-11)15(21)20(16(22)18-14)5-6-25-26(4,23)24/h7-8H,5-6H2,1-4H3
Solution Type Neat
SMILES O=C(N=C1C2=NC3=CC(C)=C(C)C=C3N1C)N(CCSS(=O)(C)=O)C2=O
Industry Type Forensics
Molecular Weight (g/mol) 394.47
Quantity 2.5 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.