missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ N2DL4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149173
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene belong to the RAET1 family, which consists of major histocompatibility complex (MHC) class I-related genes located in a cluster on chromosome 6q24.2-q25.3. This and RAET1G protein differ from other RAET1 proteins in that they have type I membrane-spanning sequences at their C termini rather than glycosylphosphatidylinositol anchor sequences. This protein functions as a ligand for NKG2D receptor, which is expressed on the surface of several types of immune cells, and is involved in innate and adaptive immune responses. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, Aug 2011].
Specifications
| N2DL4 | |
| Polyclonal | |
| Unconjugated | |
| RAET1E | |
| bA350J20.7; LETAL; Lymphocyte effector toxicity activation ligand; MGC125309; N2DL4; N2DL-4; NKG2D ligand 4; NKG2DL4; Rae-1 epsilon; RAE1 like transcript 4; RAE1epsilon; RAE-1-epsilon; RAE-1-like transcript 4; RAET1E; RAET1E2; Retinoic acid early transcript 1E; retinoic acid early-inducible protein 1-epsilon; RL-4; UL16 binding protein 4; UL16-binding protein 4; ULBP 4; ULBP4; ULBP4MGC125308; UNQ1867/PRO4303 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 135250 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8TD07 | |
| RAET1E | |
| Mouse myeloma cell line NS0-derived recombinant human ULBP-4/RAET1E, Gly30-Asp225. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction