missing translation for 'onlineSavingsMsg'
Learn More

N2-(S)-1-Ethoxycarbonyl-3-phenylpropyl-N8-trifluoroacetyl-L-lysine, TRC

Product Code. 30386197
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386197 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386197 Supplier Toronto Research Chemicals Supplier No. TRCE8916505G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material N2-(S)-1-Ethoxycarbonyl-3-phenylpropyl-N8-trifluoroacetyl-L-
CAS 116169-90-5
Molecular Formula C20 H27 F3 N2 O5
Quantity 5 g
Synonym N2-(1S-Ethoxycarbonyl-3-Phenylpropyl)-N6-Trifluoroacetyl-L-Lysine
InChI Formula InChI=1S/C20H27F3N2O5/c1-2-30-18(28)16(12-11-14-8-4-3-5-9-14)25-15(17(26)27)10-6-7-13-24-19(29)20(21,22)23/h3-5,8-9,15-16,25H,2,6-7,10-13H2,1H3,(H,24,29)(H,26,27)/t15-,16-/m0/s1
SMILES CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](CCCCNC(=O)C(F)(F)F)C(=O)O
IUPAC Name (2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid
Molecular Weight (g/mol) 432.43
Formula Weight 432.1872
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.