missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N2-[(Phenylmethoxy)carbonyl]-L-lysine, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | N2-[(Phenylmethoxy)carbonyl]-L-lysine |
| CAS | 2212-75-1 |
| Molecular Formula | C14H20N2O4 |
| Quantity | 10 g |
| Synonym | N2-Carboxy-L-lysine N-Benzyl Ester, N2-Carboxy-lysine N-Benzyl Ester, L-N2-carboxy-Lysine N2-benzyl Ester, N-alpha-Benzyloxycarbonyl-L-lysine, N-alpha-Carbobenzoxy-L-lysine, N2-Benzyloxycarbonyl-L-lysine, N2-Benzyloxycarbonyllysine, N2-Carboxylysine N2-benzyl Ester, Nalpha-(Benzyloxycarbonyl)lysine, alpha-Carbobenzoxy-L-lysine, alpha-N-Benzyloxycarbonyl-L-lysine, Z-Lys-OH |
| InChI Formula | InChI=1S/C14H20N2O4/c15-9-5-4-8-12(13(17)18)16-14(19)20-10-11-6-2-1-3-7-11/h1-3,6-7,12H,4-5,8-10,15H2,(H,16,19)(H,17,18)/t12-/m0/s1 |
| SMILES | NCCCC[C@H](NC(=O)OCc1ccccc1)C(=O)O |
| IUPAC Name | (2S)-6-amino-2-(phenylmethoxycarbonylamino)hexanoic acid |
| Molecular Weight (g/mol) | 280.32 |
| Formula Weight | 280.1423 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction