missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N2-Benzyloxycarbonyl-L-ornithine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | N2-Benzyloxycarbonyl-L-ornithine |
| CAS | 2640-58-6 |
| Molecular Formula | C13H18N2O4 |
| Quantity | 5 g |
| Synonym | N2-[(Phenylmethoxy)carbonyl]-L-ornithine, N2-Carboxyornithine N2-Benzyl Ester, Nalpha-Carbobenzoxy-L-ornithine |
| InChI Formula | InChI=1S/C13H18N2O4/c14-8-4-7-11(12(16)17)15-13(18)19-9-10-5-2-1-3-6-10/h1-3,5-6,11H,4,7-9,14H2,(H,15,18)(H,16,17)/t11-/m0/s1 |
| SMILES | NCCC[C@H](NC(=O)OCc1ccccc1)C(=O)O |
| IUPAC Name | (2S)-5-amino-2-(phenylmethoxycarbonylamino)pentanoic acid |
| Molecular Weight (g/mol) | 266.29 |
| Formula Weight | 266.1267 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction